Merck
  • Late complications of hydrogel scleral buckle implants and a technique for effective removal.
Late complications of hydrogel scleral buckle implants and a technique for effective removal.
Ophthalmic plastic and reconstructive surgery (2012-10-05)
Ayo-Lynn Richards, Dale R Meyer
ABSTRACT
The aim of this article is to describe complications of hydrogel (MIRAgel) scleral buckle implants that may appear many years after implantation, and to describe an effective technique to facilitate safe removal of these implants, which become extremely friable with time. Four case reports of patients with complications of hydrogel orbital implants and a technique for removal of the implants are described with review of the literature. The technique involves a conjunctival incision into each involved quadrant, followed by use of blunt instrumentation for "push" and a Frasier suction handpiece for "pull" to allow segmental removal. This technique limits the tendency of the implant to fragment, which has been a problem described with previous techniques. Cases typically present simulating an orbital tumor or infection, with the diagnosis not immediately apparent, as was the case in 3 of 4 cases in our series. The technique we used allowed for expeditious removal and prompt recovery in all 4 patients. Hydrogel scleral buckle implants can produce late complications that may be difficult to diagnose and manage. The "push-pull" technique described in this article facilitates effective removal, and may have implications for other types of orbital hydrogel implants as well.
Sigma-Aldrich
Poly(2-hydroxyethyl methacrylate), average Mv 20,000
Sigma-Aldrich
Poly(2-hydroxyethyl methacrylate), average Mv 300,000, crystalline
Sigma-Aldrich
Poly(2-hydroxyethyl methacrylate), average Mv 1,000,000
Sigma-Aldrich
Poly(2-hydroxyethyl methacrylate), BioReagent, powder, suitable for cell culture