Merck
  • Reuse of sewage sludge as a catalyst in ozonation--efficiency for the removal of oxalic acid and the control of bromate formation.
Reuse of sewage sludge as a catalyst in ozonation--efficiency for the removal of oxalic acid and the control of bromate formation.
Journal of hazardous materials (2012-10-02)
Gang Wen, Zhi-Hui Pan, Jun Ma, Zheng-Qian Liu, Lei Zhao, Jun-Jing Li
ABSTRACT
Sewage derived sludge is produced with an annual amount increase of 2% all over the world and it is an urgent issue to be addressed by human being. In the present study, sludge was converted into sludge-based catalyst (SBC) with ZnCl2 as activation agent and characterized by several methods (e.g., scanning electron microscope, X-ray photoelectron spectroscope and Fourier transform infrared spectroscope). Then it was used as a catalyst to enhance the removal of refractory organic matter, oxalic acid, and to control the formation of bromate (BrO3-) in bench semi-continuous ozonation experiments. The effects of various operating parameters on the control of BrO3- formation were investigated. Furthermore, the mechanism for the enhancement of organic matter removal and the control of BrO3- formation was discussed as well. Results indicate that the combination of SBC with ozone shows a strong synergistic effect, resulting in a notable improvement on oxalic acid removal. A crucial surface reaction mechanism for the enhancement of organic matter removal is proposed on the basis of negative effect of higher pH and no inhibition effect of tert-butanol. The control for BrO3- formation was demonstrated and the reason for its control in the process of O3/SBC is the combined effect of SBC reductive properties, ozone exposure decrease and hydrogen peroxide concentration increase.
Sigma-Aldrich
Zinc chloride solution, 1.9 M in 2-methyltetrahydrofuran
Sigma-Aldrich
Zinc chloride, AnhydroBeads, amorphous, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Zinc chloride, AnhydroBeads, amorphous, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Zinc chloride, anhydrous, powder, ≥99.995% trace metals basis
Sigma-Aldrich
Zinc chloride, 99.999% trace metals basis
Sigma-Aldrich
Zinc chloride, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Zinc chloride 0.1 M solution
Sigma-Aldrich
Zinc chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Zinc chloride solution, 0.5 M in THF
Sigma-Aldrich
Zinc chloride, reagent grade, ≥98%
Sigma-Aldrich
Zinc chloride, ACS reagent, ≥97%