Merck
  • Characterization of aroma compounds in Chinese bayberry (Myrica rubra Sieb. et Zucc.) by gas chromatography mass spectrometry (GC-MS) and olfactometry (GC-O).
Characterization of aroma compounds in Chinese bayberry (Myrica rubra Sieb. et Zucc.) by gas chromatography mass spectrometry (GC-MS) and olfactometry (GC-O).
Journal of food science (2012-09-27)
Wenhuai Kang, Yan Li, Yan Xu, Wenguang Jiang, Yongsheng Tao
ABSTRACT
The aroma-active compounds in Chinese bayberry were identified using gas chromatography-olfactometry (GC-O) and GC-mass spectrometry techniques. The volatile compounds were extracted using Liquid-liquid extraction, solvent-assisted flavor evaporation and headspace solid-phase microextraction (HS-SPME), respectively. On the basis of odor intensity, the most important aroma compounds in Chinese bayberry samples were caryophyllene, menthol, 4-terpineol, linalool oxide, linalool, benzyl alcohol, α-methylbenzyl alcohol, β-phenylethanol, 3-methylbutanoic acid and acetic acid, and so on. Moreover, HS-SPME technique was employed to investigate the aroma compounds among immature and mature waxberry fruits. The results showed that terpenes (for example, β-caryophyllene) was predominant and its concentration represented over 89.9% of the overall compounds, and alcohols, aldehydes, ketones, esters, acids, and others were typically present in lesser amounts. Finally, principal component analysis revealed that there was also significant difference between immature and mature waxberry fruits.
Product Description
Supelco
Linalool, analytical standard
Supelco
(−)-trans-Caryophyllene, analytical standard
Sigma-Aldrich
(−)-trans-Caryophyllene, ≥98.0% (sum of enantiomers, GC)
Sigma-Aldrich
Linalool, ≥97%, FCC, FG
Sigma-Aldrich
α-Methylbenzyl alcohol, ≥99%, FCC, FG
Sigma-Aldrich
1-Phenylethanol, 98%
Sigma-Aldrich
Linalool, 97%
Sigma-Aldrich
Benzyl alcohol, ReagentPlus®, ≥99%
Sigma-Aldrich
Benzyl alcohol, puriss. p.a., ACS reagent, ≥99.0% (GC)
Sigma-Aldrich
Benzyl alcohol, ACS reagent, ≥99.0%
Benzyl alcohol, European Pharmacopoeia (EP) Reference Standard
Supelco
Benzyl alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Benzyl alcohol, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Benzyl alcohol, natural, ≥98%, FG
Sigma-Aldrich
β-Caryophyllene, ≥80%, FCC, FG
Supelco
Benzyl alcohol, analytical standard
Supelco
(−)-Linalool, analytical standard
Sigma-Aldrich
L-Linalool, natural, ≥95%, FG
Sigma-Aldrich
Benzyl alcohol, ≥99%, FCC, FG
Sigma-Aldrich
Benzyl alcohol, anhydrous, 99.8%
Sigma-Aldrich
(−)-Linalool, ≥95.0% (sum of enantiomers, GC)