Merck
  • Antioxidant properties of diorganoyl diselenides and ditellurides: modulation by organic aryl or naphthyl moiety.
Antioxidant properties of diorganoyl diselenides and ditellurides: modulation by organic aryl or naphthyl moiety.
Molecular and cellular biochemistry (2012-09-18)
Mohammad Ibrahim, Waseem Hassan, Daiane Francine Meinerz, Matheus Dos Santos, Claudia V Klimaczewski, Anna M Deobald, Maricilia S Costa, Cristina W Nogueira, Nilda B V Barbosa, Joao B T Rocha
ABSTRACT
Diorganoyl dichalcogenide compouds can have antioxidant activity in different in vitro and in vivo models. Here, we have compared the potential antioxidant activity of 1-dinaphthyl diselenide (1-NapSe)(2), 2-dinaphthyl diselenide (2-NapSe)(2), 1-dinaphthyl distelluride (1-NapTe)(2), 2-dinaphthyl ditelluride (2-NapTe)(2) with their well-studied analogs diphenyl diselenide ((PhSe)(2)) and diphenyl telluride ((PhTe)(2)). (PhSe)(2), (PhTe)(2), and naphthalene analogs-inhibited Fe(II)-induced lipid peroxidation, catalytically decomposed hydrogen peroxide and oxidized thiols, such as dithiothreitol (DTT), Cysteine (CYS), dimercaptopropionic acid (DMPS), and thiophenol (PhSH). (PhSe)(2) was the less potent of the tested compounds against Fe(II)-induced lipid peroxidation in brain homogenates and the change in the organic moiety from an aryl to naphthyl group increased considerably the antioxidant potency of diselenide compounds. However, the change from aryl to naphthyl had little effect on the thio-peroxidase-like activity of diorganoyl dichalcogenides. These results suggest that minor changes in the organic moiety of aromatic diselenide compounds can modify profoundly their capacity to inhibit iron-induced lipid peroxidation. The pharmacological properties of organochalcogens are thought to be linked to their capacity of modulating oxidative stress. Consequently, it becomes important to explore the toxicological properties of dinaphthyl diselenides and ditellurides.
Sigma-Aldrich
Thiophenol, 97%
Sigma-Aldrich
Sodium thiophenolate, technical grade, 90%
Supelco
Thiophenol, analytical standard
Sigma-Aldrich
Thiophenol, ≥99%
Sigma-Aldrich
Diphenyl diselenide, purum, ≥97.0% (GC)
Sigma-Aldrich
Diphenyl diselenide, 98%