Merck
  • Efficient room-temperature O-silylation of alcohols using a SBA-15-supported cobalt(II) nanocatalyst.
Efficient room-temperature O-silylation of alcohols using a SBA-15-supported cobalt(II) nanocatalyst.
Chemistry & biodiversity (2012-09-15)
Fatemeh Rajabi, Rafael Luque, Juan Carlos Serrano-Ruiz
ABSTRACT
The O-silylation of OH groups of alcohols and phenols with hexamethyldisilazane (HMDS) was achieved in high-to-excellent yields using catalytic quantities of a SBA-15-supported cobalt(II) nanocatalyst (typically 0.5 mol-%) at room temperature and under solvent-free conditions. Furthermore, the heterogeneous catalyst showed an excellent durability and can be conveniently reused by filtration for at least twelve times without any noticeable loss of activity.
Product Description
Supelco
Benzyl alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Benzyl alcohol, natural, ≥98%, FG
Sigma-Aldrich
Benzyl alcohol, anhydrous, 99.8%
Sigma-Aldrich
Benzyl alcohol, ≥99%, FCC, FG
Supelco
Benzyl alcohol, analytical standard
Sigma-Aldrich
Benzyl alcohol, puriss. p.a., ACS reagent, ≥99.0% (GC)
Sigma-Aldrich
Benzyl alcohol, ReagentPlus®, ≥99%
Sigma-Aldrich
Benzyl alcohol, ACS reagent, ≥99.0%
Benzyl alcohol, European Pharmacopoeia (EP) Reference Standard