Merck
  • Effects of dexamethasone and pheniramine hydrogen maleate on stress response in patients undergoing elective laparoscopic cholecystectomy.
Effects of dexamethasone and pheniramine hydrogen maleate on stress response in patients undergoing elective laparoscopic cholecystectomy.
American journal of surgery (2012-09-05)
Kerem Karaman, Erdal Birol Bostanci, Erol Aksoy, Murat Ulas, Tuba Yigit, Mehmet Ozcan Erdemli, Ugur Ercin, Ayse Bilgihan, Gul Saydam, Musa Akoglu
ABSTRACT
Laparoscopic cholecystectomy (LC) still leads to significant postoperative nausea and vomiting (PONV) and pain. Our aim was to evaluate the efficacy of dexamethasone or pheniramine hydrogen maleate, either alone or combined, in reducing the stress response and symptoms after LC. Patients were randomly assigned to 1 of 4 groups, each consisting of 20 patients: control, dexamethasone (8 mg/2 mL), pheniramine hydrogen maleate (45.5 mg/2 mL), and the combined group. The drugs were given before anesthesia induction. C-reactive protein levels (CRP) and visual analog scale (VAS) scores were significantly less in the dexamethasone (P = .003) and combined groups (P < .001). Both dexamethasone (P < .001) and pheniramine hydrogen maleate (P = .005) significantly reduced PONV. Dexamethasone significantly reduced postoperative pain and the systemic acute-phase response, whereas these effects were only partially attained with pheniramine hydrogen maleate. Both dexamethasone and pheniramine hydrogen maleate significantly reduced PONV. An additive effect seemed to occur if these drugs were used in combination.
Product Description
Supelco
Pheniramine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Pheniramine maleate salt