Merck
  • Esterification of bio-oil from mallee (Eucalyptus loxophleba ssp. gratiae) leaves with a solid acid catalyst: Conversion of the cyclic ether and terpenoids into hydrocarbons.
Esterification of bio-oil from mallee (Eucalyptus loxophleba ssp. gratiae) leaves with a solid acid catalyst: Conversion of the cyclic ether and terpenoids into hydrocarbons.
Bioresource technology (2012-09-04)
Xun Hu, Richard Gunawan, Daniel Mourant, Yi Wang, Caroline Lievens, Weerawut Chaiwat, Liping Wu, Chun-Zhu Li
ABSTRACT
Bio-oil from pyrolysis of mallee (Eucalyptus loxophleba ssp. gratiae) leaves differs from that obtained with wood by its content of cyclic ethers, terpenoids and N-containing organic compounds. Upgrading of the leaf bio-oil in methanol with a solid acid catalyst was investigated and it was found that the N-containing organics in the bio-oil lead to deactivation of the catalyst in the initial stage of exposure and have to be removed via employing high catalyst loading to allow the occurrence of other acid-catalysed reactions. Eucalyptol, the main cyclic ether in the bio-oil, could be converted into the aromatic hydrocarbon, p-cymene, through a series of intermediates including α-terpineol, terpinolene, and α-terpinene. Various steps such as ring-opening, dehydration, isomerisation, and aromatization were involved in the conversion of eucalyptol. The terpenoids in bio-oil could also be converted into aromatic hydrocarbons that can serve as starting materials for the synthesis of fine chemicals, via the similar processes.
Product Description
Supelco
1,8-Cineole, primary reference standard
Cineole, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Eucalyptol, natural, ≥99%, FCC, FG
Supelco
Eucalyptol, analytical standard
Sigma-Aldrich
Eucalyptol, 99%
Sigma-Aldrich
Eucalyptol, tested according to Ph. Eur.