Merck
  • [Rapid determination of benzene series in seawater by gas chromatography-mass spectrometry with static headspace extraction].
[Rapid determination of benzene series in seawater by gas chromatography-mass spectrometry with static headspace extraction].
Se pu = Chinese journal of chromatography (2012-09-01)
Hongyan Bai, Bin Han, Junhui Chen, Li Zheng, Dongfang Yang, Xiaoru Wang
ABSTRACT
A method for the simultaneous determination of 13 benzene series (BTEX) in seawater using gas chromatography-mass spectrometry with static headspace extraction (HS-GC/MS) was developed. To carefully characterize the performance of this method, several factors affecting parameters were studied in detail, such as the type of column, heating procedure, equilibrium temperature, equilibrium time and the volume ratio of gas phase to liquid phase. The optimized conditions were as follows: the polar column of DB-WAX; heating procedure, 40 degrees C kept for 4 min, then raised to 120 degrees C at 10 degrees C/min, to 180 degrees C at 25 degrees C/min; equilibrium temperature, 80 degrees C; equilibrium time, 10 min; and the volume ratio of gas phase to liquid phase, 1:1. Under the optimized conditions, the linear equations were obtained in the concentration range of 0.16-320 microg/L with correlation coefficients greater than 0.999. The limits of detection (S/N = 3) were 0.019-0.033 microg/L. The recoveries at the three spiked levels of 1.6, 16 and 160 microg/L ranged from 81. 25% to 103.73% with the relative standard deviations (RSD, n=6) from 0.3% to 4.4%. The analytical results of the practical seawater samples from Shanghai Huangpu District were satisfactory. The determination of the 13 benzene series can be finished in 12 min. The method is simple, accurate, reliable, efficient and environmental-friendly.
Sigma-Aldrich
Ethylbenzene solution, suitable for NMR (reference standard), 5% in chloroform-d (99.8 atom % D), TMS 1 %, NMR tube size 5 mm × 7 in.
Sigma-Aldrich
Ethylbenzene solution, suitable for NMR (reference standard), 10% in chloroform-d (99.8 atom % D), NMR tube size 10 mm × 8 in.
Supelco
Ethylbenzene solution, suitable for NMR (reference standard), 10% in chloroform-d (99.8 atom % D), NMR tube size 5 mm × 8 in. , ultra-thin wall
Sigma-Aldrich
Ethylbenzene solution, suitable for NMR (reference standard), 0.1% in chloroform-d (99.8 atom % D), TMS 0.1 %
Sigma-Aldrich
Ethylbenzene solution, suitable for NMR (reference standard), 10% in chloroform-d (99.8 atom % D)
Sigma-Aldrich
Ethylbenzene solution, suitable for NMR (reference standard), 1% in chloroform-d (99.8 atom % D)
Sigma-Aldrich
Ethylbenzene solution, suitable for NMR (reference standard), 0.1% in chloroform-d ("100%", 99.96 atom % D)
Supelco
Ethylbenzene solution, suitable for NMR (reference standard), 0.1% in chloroform-d (99.8 atom % D), chromium(III) acetylacetonate 0.1 mg/mL, TMS 0.01 %, NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Ethylbenzene solution, suitable for NMR (reference standard), 5% in chloroform-d (99.8 atom % D), TMS 2 %, NMR tube size 5 mm × 8 in.
Supelco
Ethylbenzene solution, suitable for NMR (reference standard), 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Ethylbenzene solution, suitable for NMR (reference standard), 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 8 mm × 8 in.
Sigma-Aldrich
Ethylbenzene solution, suitable for NMR (reference standard), 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 5 mm × 8 in. , ultra-thin wall
Sigma-Aldrich
Ethylbenzene, anhydrous, 99.8%
Supelco
Ethylbenzene, analytical standard
Sigma-Aldrich
Ethylbenzene, ReagentPlus®, 99%