Merck
  • Solid phase microextraction and LC-MS/MS for the determination of paliperidone after stereoselective fungal biotransformation of risperidone.
Solid phase microextraction and LC-MS/MS for the determination of paliperidone after stereoselective fungal biotransformation of risperidone.
Analytica chimica acta (2012-08-14)
Mariana Zuccherato Bocato, Rodrigo Almeida Simões, Leandro Augusto Calixto, Cristiane Masetto de Gaitani, Mônica Tallarico Pupo, Anderson Rodrigo Moraes de Oliveira
ABSTRACT
The present work describes for the first time the use of SPME coupled to LC-MS/MS employing the polar organic mode in a stereoselective fungal biotransformation study to investigate the fungi ability to biotransform the drug risperidone into its chiral and active metabolite 9-hydroxyrisperidone (9-RispOH). The chromatographic separation was performed on a Chiralcel OJ-H column using methanol:ethanol (50:50, v/v) plus 0.2% triethylamine as the mobile phase at a flow rate of 0.8 mL min(-1). The SPME process was performed using a C18 fiber, 30 min of extraction time and 5 min of desorption time in the mobile phase. The method was completely validated and all parameters were in agreement with the literature recommendations. The Cunninghamella echinulata fungus was able to biotransform risperidone into the active metabolite, (+)-9-RispOH, resulting in 100% of enantiomeric excess. The Cunninghamella elegans fungus was also able to stereoselectively biotransform risperidone into (+)- and (-)-9-RispOH enantiomers at different rates.
Sigma-Aldrich
Paliperidone, ≥98% (HPLC)
Pricing and availability is not currently available.
Supelco
9-Hydroxyrisperidone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Pricing and availability is not currently available.