Merck
Synthesis and NMR characterization of (Z,Z,Z,Z,E,E,ω)-heptaprenol.
Journal of the American Chemical Society (2012-08-07)
Dusan Hesek, Mijoon Lee, Jaroslav Zajíček, Jed F Fisher, Shahriar Mobashery
ABSTRACT
We describe a practical, multigram synthesis of (2Z,6Z,10Z,14Z,18E,22E)-3,7,11,15,19,23,27-heptamethyl-2,6,10,14,18,22,26-octacosaheptaen-1-ol [(Z(4),E(2),ω)-heptaprenol, 4] using the nerol-derived sulfone 8 as the key intermediate. Sulfone 8 is prepared by the literature route and is converted in five additional steps (18% yield from 8) to (Z(4),E(2),ω)-heptaprenol 4. The use of Eu(hfc)(3) as an NMR shift reagent not only enabled confirmation of the structure and stereochemistry of 4, but further enabled the structural assignment to a major side product from a failed synthetic connection. The availability by this synthesis of (Z(4),E(2),ω)-heptaprenol 4 in gram quantities will enable preparative access to key reagents for the study of the biosynthesis of the bacterial cell envelope.
Sigma-Aldrich
3-Methyl-2-buten-1-ol, ≥98%, FG
Sigma-Aldrich
3-Methyl-2-buten-1-ol, 99%