Merck
  • Bulge-like asymmetric heterodye clustering in DNA duplex results in efficient quenching of background emission based on the maximized excitonic interaction.
Bulge-like asymmetric heterodye clustering in DNA duplex results in efficient quenching of background emission based on the maximized excitonic interaction.
Chemistry (Weinheim an der Bergstrasse, Germany) (2012-07-26)
Taiga Fujii, Yuichi Hara, Takuya Osawa, Hiromu Kashida, Xingguo Liang, Yasuko Yoshida, Hiroyuki Asanuma
ABSTRACT
Asymmetric dye clusters with a single fluorophore (Cy3) and multiple quenchers (4'-methylthioazobenzene-4-carboxylate, methyl red, and 4'-dimethylamino-2-nitroazobenzene-4-carboxylate) were prepared. The dye and one-to-five quenchers were tethered through D-threoninol to opposite strands of a DNA duplex. NMR analysis revealed that the clusters with a single fluorophore and two quenchers formed a sandwich-like structure (antiparallel H-aggregates). The melting temperatures of all the heteroclusters were almost the same, although structural distortion should become larger, as the number of quenchers increased. An asymmetric heterocluster of a single fluorophore and two quenchers showed larger excitonic interaction (i.e., hypochromicity of Cy3), than did a single Cy3 and a single quencher. Due to the larger exciton coupling between the dyes, the 1:2 heterocluster suppressed the background emission more efficiently than the 1:1 cluster. However, more quenchers did not enhance quenching efficiency due to the saturation of exciton coupling with two quenchers. Finally, this asymmetric 1:2 heterocluster was introduced into the stem region of a molecular beacon (MB; also known as an in-stem MB) targeting the fusion site in the L6 BCR-ABL fusion gene. With this MB design, the signal/background ratio was as high as 68 due to efficient suppression of background emission resulting from the maximized excitonic interaction.
Sigma-Aldrich
Methyl Red, ACS reagent, crystalline
Sigma-Aldrich
Methyl Red sodium salt, Crystalline
Sigma-Aldrich
Methyl Red sodium salt, ACS reagent, Dye content 95 %