MilliporeSigma
  • Enantioselective copper-catalyzed construction of aryl pyrroloindolines via an arylation-cyclization cascade.
Enantioselective copper-catalyzed construction of aryl pyrroloindolines via an arylation-cyclization cascade.
Journal of the American Chemical Society (2012-06-22)
Shaolin Zhu, David W C MacMillan
ABSTRACT
An enantioselective arylation-cyclization cascade has been accomplished using a combination of diaryliodonium salts and asymmetric copper catalysis. These mild catalytic conditions provide a new strategy for the enantioselective construction of pyrroloindolines, an important alkaloid structural motif that is commonly found among biologically active natural products.
Sigma-Aldrich
Indoline, ReagentPlus®, 99%