Merck
  • Molecular recognitive adsorption of aqueous tetramethylammonium on the organic derivative of Hiroshima University Silicate-1 with a silane coupling reagent.
Molecular recognitive adsorption of aqueous tetramethylammonium on the organic derivative of Hiroshima University Silicate-1 with a silane coupling reagent.
Chemical communications (Cambridge, England) (2012-06-12)
Yusuke Ide, Masato Torii, Nao Tsunoji, Masahiro Sadakane, Tsuneji Sano
ABSTRACT
Hiroshima University Silicate-1 (HUS-1), composed of silicate sheets with a halved sodalite cage and the interlayer tetramethylammonium (TMA) cation in the cage, was modified with dimethyldichlorosilane to form the organic derivative in which a dimethyl group was grafted onto the interlayer surface and a part of the TMA was removed, and the silylated HUS-1 effectively and selectively adsorbed TMA from water even in the presence of aqueous phenol.
Sigma-Aldrich
Tetramethylammonium chloride solution, for molecular biology
Sigma-Aldrich
Tetramethylammonium hydroxide pentahydrate, ≥95.0% (T)
Sigma-Aldrich
Tetramethylammonium hydroxide pentahydrate, ≥97%
Supelco
Tetramethylammonium bromide, suitable for ion pair chromatography, LiChropur, ≥99.0% (AT)
Sigma-Aldrich
Tetramethylammonium iodide, 99%
Sigma-Aldrich
Tetramethylammonium bromide, ACS reagent, ≥98.0%
Sigma-Aldrich
Tetramethylammonium bromide, 98%
Sigma-Aldrich
Tetramethylammonium fluoride, 97%
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 25 wt. % in H2O
Sigma-Aldrich
Tetramethylammonium chloride, reagent grade, ≥98%
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 10 wt. % in H2O
Sigma-Aldrich
Tetramethylammonium hydroxide solution, ACS reagent
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 25 wt. % in methanol
Supelco
Tetramethylammonium chloride, suitable for ion pair chromatography, LiChropur, ≥99.0% (AT)
Supelco
Tetramethylammonium sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0% (T)
Sigma-Aldrich
Tetramethylammonium chloride, BioUltra, for molecular biology, ≥99.0% (AT)