Merck
  • Nuclear translocation of DJ-1 during oxidative stress-induced neuronal cell death.
Nuclear translocation of DJ-1 during oxidative stress-induced neuronal cell death.
Free radical biology & medicine (2012-06-12)
Su-Jeong Kim, Yun-Jong Park, Ih-Yeon Hwang, Moussa B H Youdim, Kang-Sik Park, Young J Oh
ABSTRACT
Loss-of-function mutations in the PARK7/DJ-1 gene cause early onset autosomal-recessive Parkinson disease. DJ-1 has been implicated in protection of neurons from oxidative stress and in regulation of transcriptional activity. However, whether there is a relationship between the subcellular localization of DJ-1 and its function remains unknown. Therefore, we examined the subcellular localization of DJ-1 during dopaminergic neurodegeneration induced by various insults. Immunoblotting and immunocytochemistry showed that the nuclear pool of DJ-1 dramatically increased in both MN9D dopaminergic neuronal cells and primary cultures of mesencephalic dopaminergic neurons after 6-hydroxydopamine (6-OHDA) treatment. This was paralleled by a corresponding decrease in its cytosolic level, indicating drug-induced nuclear translocation of DJ-1. The same phenomenon was detected in other cell death paradigms induced by pro-oxidants including hydrogen peroxide and cupric chloride. Consequently, cotreatment with the antioxidant N-acetyl-l-cysteine blocked the translocation of DJ-1 into the nucleus. However, mutation at cysteine 106 had no effect on the translocation of DJ-1 into the nucleus, suggesting that reactive oxygen species-mediated downstream signaling and/or modifications other than oxidative modification are involved in its nuclear translocation. Ectopic expression of nucleus localization signal (NLS)-tagged DJ-1 prevented cell death from 6-OHDA. We investigated whether nuclear DJ-1 was involved in transcriptional regulation and found that DJ-1 was localized in promyelocytic leukemia bodies, and this localization increased upon 6-OHDA treatment. We also confirmed that binding of DJ-1 and promyelocytic leukemia bodies indeed increased after 6-OHDA treatment. Consequently, expression levels of acetylated p53 and PUMA were downregulated in cells overexpressing DJ-1 or NLS-tagged DJ-1. Taken together, our data suggest that nuclear translocation of DJ-1 may protect neurons from cell death after oxidative stress.
Sigma-Aldrich
Copper(II) chloride, powder, 99%
Sigma-Aldrich
Copper(II) chloride, 99.999% trace metals basis
Sigma-Aldrich
Copper(II) chloride, anhydrous, powder, ≥99.995% trace metals basis
Sigma-Aldrich
Copper(II) chloride, 97%
Sigma-Aldrich
Copper(II) chloride dihydrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Copper(II) chloride dihydrate, 99.999%
Sigma-Aldrich
Copper(II) chloride dihydrate, ≥99.95% trace metals basis
Sigma-Aldrich
Copper(II) chloride dihydrate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Copper(II) chloride dihydrate, reagent grade