Merck
  • Improved protocol for indoline synthesis via palladium-catalyzed intramolecular C(sp2)-H amination.
Improved protocol for indoline synthesis via palladium-catalyzed intramolecular C(sp2)-H amination.
Organic letters (2012-06-08)
Gang He, Chengxi Lu, Yingsheng Zhao, William A Nack, Gong Chen
ABSTRACT
An efficient method has been developed for the synthesis of indoline compounds from picolinamide (PA)-protected β-arylethylamine substrates via palladium-catalyzed intramolecular amination of ortho-C(sp(2))-H bonds. These reactions feature high efficiency, low catalyst loadings, mild operating conditions, and the use of inexpensive reagents.
Sigma-Aldrich
Indoline, ReagentPlus®, 99%