Merck
  • [Determination of 9 residual acrylic monomers in acrylic resins by gas chromatography-mass spectrometry coupled with microwave assisted extraction].
[Determination of 9 residual acrylic monomers in acrylic resins by gas chromatography-mass spectrometry coupled with microwave assisted extraction].
Se pu = Chinese journal of chromatography (2012-06-07)
Ying Lai, Rui Lin, Luxin Cai, Xiuxiu Ge, Changchun Huang
ABSTRACT
A reliable gas chromatography-mass spectrometry (GC-MS) method was developed for the determination of 9 residual acrylic monomers (methyl acrylate, ethyl acrylate, methyl methacrylate, ethyl methacrylate, n-butyl acrylate, butyl methacrylate, styrene, acrylic acid and methacrylic acid) in acrylic resins. Solid resin was precipitated with methanol after microwave assisted extraction with ethyl acetate for 30 min, and liquid resin was diluted with methanol directly. The nine acrylic monomers got a good separation within 20 min on a DB-WAX column. The limits of quantification (LOQs, S/N = 10) of the method were in the range of 1-10 mg/kg for liquid resin and 3-50 mg/kg for solid resin. The calibration curves were linear within 1-500 mg/L range with correlation coefficients above 0. 995. The recoveries ranged from 84.4% to 108.6% at five spiked levels. The sensitivity, recovery and selectivity of the method can fully meet the requirements of practical work.
Sigma-Aldrich
Ethyl methacrylate, contains 15-20 ppm monomethyl ether hydroquinone as inhibitor, 99%
Sigma-Aldrich
Methyl acrylate, 99%, contains ≤100 ppm monomethyl ether hydroquinone as inhibitor
Supelco
Ethyl acrylate, analytical standard
Sigma-Aldrich
Ethyl acrylate, contains 10-20 ppm MEHQ as inhibitor, 99%
Supelco
Methyl acrylate, analytical standard
Sigma-Aldrich
Ethyl acrylate, ≥99.5%, stabilized