Merck
  • Idebenone-loaded solid lipid nanoparticles for drug delivery to the skin: in vitro evaluation.
Idebenone-loaded solid lipid nanoparticles for drug delivery to the skin: in vitro evaluation.
International journal of pharmaceutics (2012-06-05)
Lucia Montenegro, Chiara Sinico, Ines Castangia, Claudia Carbone, Giovanni Puglisi
ABSTRACT
Idebenone (IDE), a synthetic derivative of ubiquinone, shows a potent antioxidant activity that could be beneficial in the treatment of skin oxidative damages. In this work, the feasibility of targeting IDE into the upper layers of the skin by topical application of IDE-loaded solid lipid nanoparticles (SLN) was evaluated. SLN loading different amounts of IDE were prepared by the phase inversion temperature method using cetyl palmitate as solid lipid and three different non-ionic surfactants: ceteth-20, isoceteth-20 and oleth-20. All IDE loaded SLN showed a mean particle size in the range of 30-49 nm and a single peak in size distribution. In vitro permeation/penetration experiments were performed on pig skin using Franz-type diffusion cells. IDE penetration into the different skin layers depended on the type of SLN used while no IDE permeation occurred from all the SLN under investigation. The highest IDE content was found in the epidermis when SLN contained ceteth-20 or isoceteth-20 as surfactant while IDE distribution into the upper skin layers depended on the amount of IDE loaded when oleth-20 was used as surfactant. These results suggest that the SLN tested could be an interesting carrier for IDE targeting to the upper skin layers.
Sigma-Aldrich
Idebenone, ≥98% (HPLC)
Sigma-Aldrich
Brij® 93, average Mn ~357
Sigma-Aldrich
SP Brij® C2 MBAL-SO-(SG), average Mn ~330
Sigma-Aldrich
Brij® 58, average Mn ~1124
Sigma-Aldrich
Brij® O10
Sigma-Aldrich
ECO BRIJ® C10
Sigma-Aldrich
ECO BRIJ® O20, average Mn ~1,150
Sigma-Aldrich
ECO BRIJ® O10
Sigma-Aldrich
BRIJ® O20, average Mn ~1,150
Sigma-Aldrich
Brij® C10, average Mn ~683