Merck
  • Complications of modeling glycosylation reactions: can the anomeric conformation of a donor determine the glycopyranosyl oxacarbenium ring conformation?
Complications of modeling glycosylation reactions: can the anomeric conformation of a donor determine the glycopyranosyl oxacarbenium ring conformation?
Carbohydrate research (2012-05-01)
Dennis M Whitfield
ABSTRACT
That the ring conformation of glycopyranosyl oxacarbenium ions can influence the stereochemical outcome of glycosylation reactions has been postulated for some time. Some new ionization calculations show that the ultimate conformation (4)H(3) or (5)S(1) of D-glucopyranosyl oxacarbenium ions depends on the initial ϕ(H) (CH-1-C-1-S(+)-SCH(3)) conformation of anomeric thiosulfonium ions. Evidence is also presented that nucleophile:electrophile hydrogen bonded complexes, 1,6-anhydro-carbenium ions and electron rich carbon nucleophile:oxacarbenium ion complexes are all probably artifacts of neglecting counter ions or nucleophiles in the DFT calculation. All three cationic species are likely important for glycosylation reaction side reactions but not as productive species.
Sigma-Aldrich
Lithium trifluoromethanesulfonate, 99.995% trace metals basis
Sigma-Aldrich
Zinc trifluoromethanesulfonate, 98%
Sigma-Aldrich
Trifluoromethanesulfonic acid, reagent grade, 98%
Sigma-Aldrich
Lithium trifluoromethanesulfonate, 96%
Sigma-Aldrich
Trifluoromethanesulfonic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Lanthanum(III) trifluoromethanesulfonate, 99.999% trace metals basis
Sigma-Aldrich
Silver trifluoromethanesulfonate, ≥99%
Sigma-Aldrich
Silver trifluoromethanesulfonate, ≥99.95% trace metals basis
Sigma-Aldrich
Silver trifluoromethanesulfonate, purum, ≥98.0% (Ag)
Sigma-Aldrich
Aluminum trifluoromethanesulfonate, 99.9% trace metals basis