Merck
  • Prognosis after high-dose chemotherapy followed by autologous stem-cell transplantation as first-line treatment in primary CNS lymphoma--a long-term follow-up study.
Prognosis after high-dose chemotherapy followed by autologous stem-cell transplantation as first-line treatment in primary CNS lymphoma--a long-term follow-up study.
Annals of oncology : official journal of the European Society for Medical Oncology (2012-04-05)
B Kasenda, E Schorb, K Fritsch, J Finke, G Illerhaus
ABSTRACT
High-dose chemotherapy followed by autologous stem-cell transplantation (HCT-ASCT) is a promising approach in eligible patients with primary central nervous system lymphoma (PCNSL). We report long-term data of patients who were treated according to HCT-ASCT containing protocols. We analyzed survival and relapse rates in 43 (<67 years) immunocompetent patients with newly diagnosed PCNSL being treated according to two different high-dose methotrexate-based protocols followed by high-dose carmustine/thiotepa (BCNU/TT) plus ASCT (±whole brain irradiation). Analysis was conducted for all patients (intention-to-treat) and those patients who actually received HCT-ASCT (per-protocol). Thirty-four patients achieved complete remission, of those 12 relapsed (35%), while 6 of them relapsed 5 years after diagnosis. After a median follow-up of 120 months, median overall survival (OS) was reached after 104 months. Two- and 5-year OS was 81% and 70% and 2- and 5-year event-free survival (EFS) was 81% and 67%, respectively. In per-protocol analysis (N = 34), 5-year OS and EFS was 82% and 79%, respectively. HCT-ASCT associated related mortality was not observed. Sequential high-dose MTX containing chemotherapy followed by high-dose carmustine/thiotepa plus ASCT (±whole brain irradiation) is safe and leads to high survival rates in eligible patients with newly diagnosed PCNSL.
Sigma-Aldrich
Thio-TEPA
Pricing and availability is not currently available.