Merck
  • Design, synthesis, and structure-activity correlations of novel dibenzo[b,d]furan, dibenzo[b,d]thiophene, and N-methylcarbazole clubbed 1,2,3-triazoles as potent inhibitors of Mycobacterium tuberculosis.
Design, synthesis, and structure-activity correlations of novel dibenzo[b,d]furan, dibenzo[b,d]thiophene, and N-methylcarbazole clubbed 1,2,3-triazoles as potent inhibitors of Mycobacterium tuberculosis.
Journal of medicinal chemistry (2012-03-28)
Santhosh Reddy Patpi, Lokesh Pulipati, Perumal Yogeeswari, Dharmarajan Sriram, Nishant Jain, Balasubramanian Sridhar, Ramalinga Murthy, T Anjana Devi, Shasi Vardhan Kalivendi, Srinivas Kantevari
ABSTRACT
A molecular hybridization approach is an emerging structural modification tool to design new molecules with improved pharmacophoric properties. In this study, 1,2,3-triazole-based Mycobacterium tuberculosis inhibitors and synthetic and natural product-based tricyclic (carbazole, dibenzo[b,d]furan, and dibenzo[b,d]thiophene) antimycobacterial agents were integrated in one molecular platform to prepare various novel clubbed 1,2,3-triazole hybrids using click chemistry. Structure-activity correlations and in vitro activity against M. tuberculosis strain H37Rv of new analogues revealed the order: dibenzo[b,d]thiophene > dibenzo[b,d]furan > 9-methyl-9H-carbazole series. Two of the most potent M. tuberculosis inhibitors 13h and 13q with MIC = 0.78 μg/mL (∼1.9 μM) displayed a low cytotoxicity and high selectivity index (50-255) against four different human cancer cell lines. These results together provided the potential importance of molecular hybridization and the development of triazole clubbed dibenzo[b,d]thiophene-based lead candidates to treat mycobacterial infections.
Product Description
Supelco
Dibenzothiophene, analytical standard
Sigma-Aldrich
Dibenzothiophene, ≥99%
Sigma-Aldrich
Dibenzothiophene, 98%