Merck
  • Visible light mediated homo- and heterocoupling of benzyl alcohols and benzyl amines on polycrystalline cadmium sulfide.
Visible light mediated homo- and heterocoupling of benzyl alcohols and benzyl amines on polycrystalline cadmium sulfide.
Organic & biomolecular chemistry (2012-03-27)
Tatiana Mitkina, Christoph Stanglmair, Wolfgang Setzer, Michael Gruber, Horst Kisch, Burkhard König
ABSTRACT
The oxidative coupling of sp(3) hybridized carbon atoms by photocatalysis is a valuable synthetic method as stoichiometric oxidation reagents can be avoided and dihydrogen is the only byproduct of the reaction. Cadmium sulfide, a readily available semiconductor, was used as a visible light heterogeneous photocatalyst for the oxidative coupling of benzyl alcohols and benzyl amines by irradiation with blue light. Depending on the structure of the starting material, good to excellent yields of homocoupling products were obtained as mixtures of diastereomers. Cross-coupling between benzyl alcohols and benzyl amines gave product mixtures, but was selective for the coupling of tetrahydroisoquinolines to nitromethane. The results demonstrate that CdS is a suitable visible light photocatalyst for oxidative bond formation under anaerobic conditions.
Sigma-Aldrich
Benzyl alcohol, puriss. p.a., ACS reagent, ≥99.0% (GC)
Sigma-Aldrich
Benzyl alcohol, ReagentPlus®, ≥99%
Supelco
Benzyl alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Benzyl alcohol, anhydrous, 99.8%
Sigma-Aldrich
Benzyl alcohol, ≥99%, FCC, FG
Supelco
Benzyl alcohol, analytical standard
Sigma-Aldrich
Benzyl alcohol, natural, ≥98%, FG
Sigma-Aldrich
Benzyl alcohol, ACS reagent, ≥99.0%
Benzyl alcohol, European Pharmacopoeia (EP) Reference Standard