Merck
  • Electrochemical synthesis of adiponitrile from the renewable raw material glutamic acid.
Electrochemical synthesis of adiponitrile from the renewable raw material glutamic acid.
ChemSusChem (2012-03-24)
Jian-Jun Dai, Yao-Bing Huang, Chi Fang, Qing-Xiang Guo, Yao Fu
ABSTRACT
Current affairs: Adiponitrile, used to produce nylon 6.6, is prepared from the renewable compound glutamic acid by an electrochemical route, involving electro-oxidative decarboxylation and Kolbe coupling reactions. The new route is an example of the use of glutamic acid as a versatile substrate in the transformation of biomass into chemicals. Also, it highlights the use of electrochemical methods in biomass conversion.
Sigma-Aldrich
Adiponitrile, 99%