- Synthesis of dendritic platinum nanoparticles/lucigenin/reduced graphene oxide hybrid with chemiluminescence activity.
Synthesis of dendritic platinum nanoparticles/lucigenin/reduced graphene oxide hybrid with chemiluminescence activity.
Chemistry (Weinheim an der Bergstrasse, Germany) (2012-03-16)
Yi He, Hua Cui
PMID22419477
ABSTRACT
Multifunctional hybrid: A dendritic platinum nanoparticle/lucigenin/reduced graphene oxide (RGO) hybrid with chemiluminescence (CL) activity was prepared for the first time by using lucigenin as a linker through simultaneous reduction of H(2) PtCl(4) and a lucigenin-functionalized graphene oxide composite by sodium borohydride (see scheme). The hybrid may have potential applications as a CL sensor, in catalysis, energy conversion, and opto-electronic systems.