Merck
  • Clinical evidence for the superior efficacy of a dentifrice containing 8.0% arginine and calcium carbonate in providing instant and lasting relief of dentin hypersensitivity.
Clinical evidence for the superior efficacy of a dentifrice containing 8.0% arginine and calcium carbonate in providing instant and lasting relief of dentin hypersensitivity.
The Journal of clinical dentistry (2011-01-01)
D Cummins
ABSTRACT
This paper briefly discusses recent scientific and clinical research validating the effectiveness of a toothpaste containing 8.0% arginine and calcium carbonate, known as Pro-Argin technology, including clinical evidence for the superior efficacy of this toothpaste versus a potassium-based desensitizing toothpaste. It also introduces new clinical data which prove that a toothpaste containing 8.0% arginine and calcium carbonate delivers superior instant and lasting relief of dentin hypersensitivity compared to a toothpaste containing 8% strontium acetate.
Sigma-Aldrich
Strontium acetate
Sigma-Aldrich
Strontium acetate, 99.95% trace metals basis