Merck
  • Ultrasound assisted synthesis of tetrahydrobenzo[c]xanthene-11-ones using CAN as catalyst.
Ultrasound assisted synthesis of tetrahydrobenzo[c]xanthene-11-ones using CAN as catalyst.
Ultrasonics sonochemistry (2012-03-09)
S Sudha, M A Pasha
ABSTRACT
Tetrahydrobenzo[c]xanthenes-11-ones was synthesized by a three component reaction of α-naphthol, aromatic aldehyde and dimedone. Ceric ammonium nitrate acts as a suitable eco-friendly catalyst for this method. Shorter reaction duration, mild reaction condition and low cost make this protocol more effective.
Sigma-Aldrich
Ammonium cerium(IV) nitrate, puriss. p.a., ACS reagent, ≥98.5% (RT)
Sigma-Aldrich
Ammonium cerium(IV) nitrate, ACS reagent, ≥98.5%
Sigma-Aldrich
Ammonium cerium(IV) nitrate, ≥98% (titration)
Sigma-Aldrich
Ammonium cerium(IV) nitrate, ≥99.99% trace metals basis