Merck
  • Functional and genetic characterization of the tap efflux pump in Mycobacterium bovis BCG.
Functional and genetic characterization of the tap efflux pump in Mycobacterium bovis BCG.
Antimicrobial agents and chemotherapy (2012-01-11)
Santiago Ramón-García, Virginie Mick, Elisa Dainese, Carlos Martín, Charles J Thompson, Edda De Rossi, Riccardo Manganelli, José A Aínsa
ABSTRACT
Efflux pumps extrude a wide variety of chemically unrelated compounds conferring multidrug resistance and participating in numerous physiological processes. Mycobacterium tuberculosis possesses many efflux pumps, and their roles in drug resistance and physiology are actively investigated. In this work we found that tap mutant cells showed changes in morphology and a progressive loss of viability upon subcultivation in liquid medium. Transcriptome analysis in Mycobacterium bovis BCG revealed that disruption of the Rv1258c gene, encoding the Tap efflux pump, led to an extensive change in gene expression patterns during stationary phase, with no changes during exponential growth. In stationary phase, Tap inactivation triggered a general stress response and led to a general repression of genes involved in cell wall biosynthesis, in particular the formation of the peptidoglycan; this suggested the accumulation of an unknown Tap substrate that reaches toxic concentrations during stationary phase. We also found that both disruption and overexpression of tap altered susceptibility to many clinically approved antibiotics in M. bovis BCG. Acriflavine and tetracycline accumulation assays and carbonyl cyanide m-chlorophenylhydrazone (CCCP) potentiation experiments demonstrated that this phenotype was due to an active efflux mechanism. These findings emphasize the important role of the Tap efflux pump in bacterial physiology and intrinsic drug resistance.
Sigma-Aldrich
Auramine O, Dye content 85 %, certified by the Biological Stain Commission
Sigma-Aldrich
Acriflavine hydrochloride, fluorescent dye
Sigma-Aldrich
Auramine O, Dye content ≥80 %, certified by the Biological Stain Commission
Sigma-Aldrich
Acriflavine, BioReagent, suitable for fluorescence, ≥90% (AT)
Sigma-Aldrich
Acriflavine, fluorescent label