Merck
  • Study of an aquifer contaminated by ethyl tert-butyl ether (ETBE): site characterization and on-site bioremediation.
Study of an aquifer contaminated by ethyl tert-butyl ether (ETBE): site characterization and on-site bioremediation.
Journal of hazardous materials (2011-12-20)
Françoise Fayolle-Guichard, Jonathan Durand, Mathilde Cheucle, Mònica Rosell, Rory Julien Michelland, Jean-Philippe Tracol, Françoise Le Roux, Geneviève Grundman, Olivier Atteia, Hans H Richnow, Alain Dumestre, Yves Benoit
ABSTRACT
Ethyl tert-butyl ether (ETBE) was detected at high concentration (300mgL(-1)) in the groundwater below a gas-station. No significant carbon neither hydrogen isotopic fractionation of ETBE was detected along the plume. ETBE and BTEX biodegradation capacities of the indigenous microflora Pz1-ETBE and of a culture (MC-IFP) composed of Rhodococcus wratislaviensis IFP 2016, Rhodococcus aetherivorans IFP 2017 and Aquincola tertiaricarbonis IFP 2003 showed that ETBE and BTEX degradation rates were in the same range (ETBE: 0.91 and 0.83 mg L(-1)h(-1) and BTEX: 0.64 and 0.82 mg L(-1)h(-1), respectively) but tert-butanol (TBA) accumulated transiently at a high level using Pz1-ETBE (74 mg L(-1)). An on-site pilot plant (2m(3)) filled with polluted groundwater and inoculated by MC-IFP, successfully degraded four successive additions of ETBE and gasoline. However, an insignificant ETBE isotopic fractionation was also accompanying this decrease which suggested the involvement of low fractionating-strains using EthB enzymes, but required of additional proofs. The ethB gene encoding a cytochrome P450 involved in ETBE biodegradation (present in R. aetherivorans IFP 2017) was monitored by quantitative real-time polymerase chain reaction (q-PCR) on DNA extracted from water sampled in the pilot plant which yield up to 5×10(6) copies of ethB gene per L(-1).
Product Description
Supelco
tert-Butyl ethyl ether, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
tert-Butyl ethyl ether, purum, ≥97.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
tert-Butyl ethyl ether, 99%
Pricing and availability is not currently available.