Merck
  • Chirality sensing by a fluorescent binaphthocrown ether-polythiophene conjugate.
Chirality sensing by a fluorescent binaphthocrown ether-polythiophene conjugate.
Chemical communications (Cambridge, England) (2011-12-15)
Gaku Fukuhara, Yoshihisa Inoue
ABSTRACT
Chiral recognition abilities of the title host for (R)- and (S)-α-methyl-4-nitrobenzylamine were examined in the ground and excited states to give a relative affinity (K(R)/K(S)) of 2.16 by spectral titration and a relative rate constant (k(R)/k(S)) of 2.23 by fluorescence quenching, revealing that the quenching process is static and not enantiodifferentiating.
Sigma-Aldrich
(S)-α-Methyl-4-nitrobenzylamine hydrochloride, 97%
Sigma-Aldrich
4-Nitrobenzylamine hydrochloride, 97%