Merck
  • Study on the metabolic mechanism of chiral inversion of S-mandelic acid in vitro.
Study on the metabolic mechanism of chiral inversion of S-mandelic acid in vitro.
Chirality (2011-12-06)
Ling-Bo Gao, Jin-Zhao Wang, Tong-Wei Yao, Su Zeng
ABSTRACT
Mandelic acid (MA) is generally used as a biological indicator of occupational exposure to styrene, which is classified as a class of hazardous environmental pollutants. It was found to undergo one-directional chiral inversion (S-MA to R-MA) in Wistar and Sprague-Dawley rats in vivo. This study was aimed to explore the metabolic mechanism of chiral inversion of S-MA in vitro. S-MA was converted to R-MA in rat hepatocytes, whereas MA enantiomers remained unchanged in acidic and neutral phosphate buffers, HepG2 cells, and intestinal flora. In addition, the synthesized S-MA-CoA thioester was rapidly racemized and hydrolyzed to R-MA by rat liver homogenate and S9, cytosolic and mitochondrial fractions. The data suggest that chiral inversion of S-MA may involve the hydrolysis of S-MA-CoA, and its metabolic mechanism could be the same as that of 2-arylpropionic acid (2-APA) drugs.
Product Description
Supelco
DL-Mandelic acid, analytical standard
Sigma-Aldrich
(R)-(−)-Mandelic acid, 98%
Sigma-Aldrich
Mandelic acid, 99%
Sigma-Aldrich
(R)-(−)-Mandelic acid, ChiPros®, produced by BASF, 99%
Sigma-Aldrich
(R)-(−)-Mandelic acid, ReagentPlus®, ≥99%
Sertraline impurity E, European Pharmacopoeia (EP) Reference Standard