Merck
  • Chlorpromazine quantification in human plasma by UPLC-electrospray ionization tandem mass spectrometry. Application to a comparative pharmacokinetic study.
Chlorpromazine quantification in human plasma by UPLC-electrospray ionization tandem mass spectrometry. Application to a comparative pharmacokinetic study.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2011-11-15)
Ney Carter Borges, Vinicius Marcondes Rezende, Jose Marcos Santana, Ricardo Pereira Moreira, Roberto Fernandes Moreira, Patrícia Moreno, Diego Carter Borges, José Luiz Donato, Ronilson Agnaldo Moreno
ABSTRACT
In the present study a method to quantify chlorpromazine in human plasma using cyclobenzaprine as the internal standard (IS) is described. The analyte and the IS were extracted from human plasma by a liquid-liquid extraction with diethyl ether/dichloromethane (70/30, v/v) and analyzed by an ultra performance liquid chromatography (UPLC) coupled to an electrospray tandem triple quadrupole mass spectrometer in positive mode (UPLC-ES(+)-MS/MS). Chromatography was performed isocratically on an Aquity UPLC BEH C18 1.7 μm (50 mm × 2.1 mm i.d.) operating at 40°C. The mobile phase was a mixture of 65% water+1% formic acid and 35% of acetonitrile at a flow-rate of 0.5 mL/min. The lowest concentration quantified was 0.5 ng/mL and a linear calibration curve over the range 0.5-200 ng/mL was obtained, showing intra-assay precisions from 2.4 to 5.8%, and inter-assay precisions from 3.6 to 9.9%. The intra-assay accuracies ranged from 96.9 to 102.5%, while the inter-assay accuracies ranged from 94.1 to 100.3%. This analytical method was applied in a relative bioavailability study in order to compare a test chlorpromazine 100 mg simple dose formulation versus a reference in 57 volunteers of both sexes. The study was conducted in an open randomized two-period crossover design and with a fourteen days washout period. Plasma samples were obtained over a 144-h interval. Since the 90% CI for both C(max), AUC(last) and AUC(0-inf) were within the 80-125% interval proposed by the Food and Drug Administration and ANVISA, it was concluded that chlorpromazine 100 mg/dose was bioequivalent to the reference formulation, according to both the rate and extent of absorption.
Product Description
Supelco
Cyclobenzaprine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Cyclobenzaprine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Cyclobenzaprine hydrochloride