Merck
  • Integrative and intermediate self-assembly of multi-walled hybrid nanotubes for catanionic biomimetics.
Integrative and intermediate self-assembly of multi-walled hybrid nanotubes for catanionic biomimetics.
Chemical communications (Cambridge, England) (2011-10-25)
Fenglin Liu, Wenpei Kang, Chenhao Zhao, Yunlan Su, Dujin Wang, Qiang Shen
ABSTRACT
Hollow cylindrical multi-walled hybrid nanotubes go through dynamic growth and subsequent disappearance during the biomimetic fabrication of hexagonal calcite platelets, simulating the in vivo purpose-driven self-assembly of tubular plasma-membrane calcium-ion channels for biomaterials to adapt, respond and repair.
Sigma-Aldrich
Ammonium lauryl sulfate solution, ~30% in H2O (T)
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)
Sigma-Aldrich
Diethyl carbonate, anhydrous, ≥99%
Sigma-Aldrich
Lithium dodecyl sulfate, BioReagent, for molecular biology, suitable for electrophoresis
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)
Sigma-Aldrich
Lithium dodecyl sulfate, BioXtra, ≥98.5% (GC)
Sigma-Aldrich
Lithium dodecyl sulfate, ≥98.5% (GC)
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)
Sigma-Aldrich
Ammonium lauryl sulfate solution, ~30% in H2O
Sigma-Aldrich
Diethyl carbonate, 99%