Merck
  • Robustness of an aerobic metabolically vinyl chloride degrading bacterial enrichment culture.
Robustness of an aerobic metabolically vinyl chloride degrading bacterial enrichment culture.
Water science and technology : a journal of the International Association on Water Pollution Research (2011-10-25)
He-Ping Zhao, Kathrin R Schmidt, Svenja Lohner, Andreas Tiehm
ABSTRACT
Degradation of the lower chlorinated ethenes is crucial to the application of natural attenuation or in situ bioremediation on chlorinated ethene contaminated sites. Recently, within mixtures of several chloroethenes as they can occur in contaminated groundwater inhibiting effects on aerobic chloroethene degradation have been shown. The current study demonstrated that metabolic vinyl chloride (VC) degradation by an enrichment culture originating from groundwater was not affected by an equimolar concentration (50 μM) of cis-1,2-dichloroethene (cDCE). Only cDCE concentrations at a ratio of 2.4:1 (initial cDCE to VC concentration) caused minor inhibition of VC degradation. Furthermore, the degradation of VC was not affected by the presence of trans-1,2-dichloroethene (tDCE), 1,1-dichloroethene (1,1-DCE), trichloroethene (TCE), and tetrachloroethene (PCE) in equimolar concentrations (50 μM). Only cDCE and tDCE were cometabolically degraded in small amounts. The VC-degrading culture demonstrated a broad pH tolerance from 5 to 9 with an optimum between 6 and 7. Results also showed that the culture could degrade VC concentrations up to 1,800 μM (110 mg/L).
Product Description
Supelco
cis-1,2-Dichloroethene solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
cis-1,2-Dichloroethylene, 97%
Sigma-Aldrich
trans-1,2-Dichloroethylene, 98%
Supelco
trans-1,2-Dichloroethylene, analytical standard
Sigma-Aldrich
1,2-Dichloroethylene, mixture of cis and trans, 98%
Supelco
cis-1,2-Dichloroethene, analytical standard