Merck
  • Dielectric properties of lead-free BZT-KNN perovskite ceramics for energy storage.
Dielectric properties of lead-free BZT-KNN perovskite ceramics for energy storage.
ChemSusChem (2011-10-14)
Dong-Yun Gui, Han-Xing Liu, Hua Hao, Yue Sun, Ming-He Cao, Zhi-Yong Yu
ABSTRACT
Lead-free (1-x)Ba(Zr₀.₁₅Ti₀.₈₅)O₃-x(K₀.₅Na₀.₅)NbO₃ ; x=0-0.05) (BZT-KNN) perovskite ceramics, a materials with potential applications for energy storage, are investigated. The samples are prepared by a solid-state reaction method. Powder X-ray diffraction (XRD) and scanning electron microscopy (SEM) are used to study the microstructure of the samples. Their dielectric properties and impedance spectra are reported as functions of temperature and frequency. The addition of 1 mol % (K₀.₅Na₀.₅)NbO₃ to Ba(Zr₀.₁₅Ti₀.₈₅)O₃ improves the dielectric constant and enhances its diffuseness in a wide temperature range. The small amount of (K₀.₅Na₀.₅)NbO₃ is found to markedly affect the microstructure of the Ba(Zr₀.₁₅Ti₀.₈₅)O₃ ceramic (grain size and other characteristics) without changing the phase or crystal symmetry. In addition, we report that fine substructures in the grains, so-called sheet structures, are responsible for the dielectric properties (both diffuseness and dielectric constant) of (1-x)Ba(Zr₀.₁₅Ti₀.₈₅)O₃-x(K₀.₅Na₀.₅)NbO₃ (x=0-0.03; especially x=0.01) ceramics.
Sigma-Aldrich
Niobium(V) oxide, 99.99% trace metals basis
Sigma-Aldrich
Niobium(V) oxide, −325 mesh, 99.9% trace metals basis
Sigma-Aldrich
Barium carbonate, ACS reagent, ≥99%
Sigma-Aldrich
Potassium carbonate, ReagentPlus®, 99%
Sigma-Aldrich
Potassium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium carbonate, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium carbonate, reagent grade, ≥98%, powder, −325 mesh
Sigma-Aldrich
Potassium carbonate, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Potassium carbonate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Barium carbonate, 99.999% trace metals basis
Sigma-Aldrich
Barium carbonate, 99.98% trace metals basis
Sigma-Aldrich
Potassium carbonate, 99.995% trace metals basis
Sigma-Aldrich
Potassium carbonate, meets USP testing specifications
Sigma-Aldrich
Potassium carbonate, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)