Merck
  • Ketogenic diet-induced peroxisome proliferator-activated receptor-γ activation decreases neuroinflammation in the mouse hippocampus after kainic acid-induced seizures.
Ketogenic diet-induced peroxisome proliferator-activated receptor-γ activation decreases neuroinflammation in the mouse hippocampus after kainic acid-induced seizures.
Experimental neurology (2011-09-24)
Eun Ae Jeong, Byeong Tak Jeon, Hyun Joo Shin, Nayoung Kim, Dong Hoon Lee, Hyun Joon Kim, Sang Soo Kang, Gyeong Jae Cho, Wan Sung Choi, Gu Seob Roh
ABSTRACT
Similar to fasting, the ketogenic diet (KD) has anti-inflammatory effects and protects against excitotoxicity-mediated neuronal cell death. Recent studies have shown that peroxisome proliferator-activated receptor (PPAR)γ has anti-inflammatory effects in seizure animal models. However, the exact mechanisms underlying the anti-inflammatory effects of the KD have not been determined for seizures. Here we investigated the effect of the KD and acetoacetate (AA) on neuroinflammation in a seizure animal model and glutamate-treated HT22 cells, respectively. Mice were fed the KD for 4 weeks and sacrificed 2 or 6h after KA injection. The KD reduced hippocampal tumor necrosis factor alpha (TNF-α) levels and nuclear factor (NF)-κB translocation into the nucleus 2h after KA treatment. KD-induced PPARγ activation was decreased by KA in neurons as assessed by western blotting and immunofluorescence. Finally, the KD inhibited cyclooxygenase (COX)-2 and microsomal prostaglandin E(2) synthase-1 (mPGES-1) expression in the hippocampus 6h after KA treatment. AA treatment also protected against glutamate-induced cell death in HT22 cells by reducing TNF-α and PPARγ-mediated COX-2 expression. Thus, the KD may inhibit neuroinflammation by suppressing a COX-2-dependent pathway via activation of PPARγ by the KD or AA.
Sigma-Aldrich
Lithium acetoacetate, ≥90% (HPLC)