Merck
  • Clinical inquiry: what's the best way to relieve mastitis in breastfeeding mothers?
Clinical inquiry: what's the best way to relieve mastitis in breastfeeding mothers?
The Journal of family practice (2011-09-09)
Aurelie Cabou, Sarah Babineau, Leilani St Anna
ABSTRACT
Frequent breast emptying helps both infectious and noninfectious mastitis. Antibiotics may be useful for women with positive milk cultures, but their utility for treating undifferentiated mastitis is unknown. Consider prescribing antibiotics for women whose mastitis symptoms don' t improve after 12 to 24 hours of frequent breast emptying.
Product Description
USP
Cephradine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Cephradine, ≥90.0% (Cephradine, HPLC)