Merck
  • Determination and study on dissipation of 1-naphthylacetic acid in garlic and soil using high performance liquid chromatography-tandem mass spectrometry.
Determination and study on dissipation of 1-naphthylacetic acid in garlic and soil using high performance liquid chromatography-tandem mass spectrometry.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2011-08-26)
Wenbi Guan, Pengjun Xu, Kai Wang, Yue Song, Hongyan Zhang
ABSTRACT
A simple, quick and reliable analytical method for the determination of 1-naphthylacetic acid in garlic and soil has been developed in this study. The residual levels and dissipation rates of 1-naphthylacetic acid in garlic and soil were determined by high performance liquid chromatography-tandem mass spectroscopy (HPLC-MS/MS). The limit of quantification (LOQ) of the developed method was 0.005 mg/kg. The half-lives of 1-naphthylacetic acid in garlic plants and soil were 0.80-1.4 days and 0.94-2.0 days, respectively. The final residues of 1-naphthylacetic acid in garlic, garlic sprout and soil could not be detected and were all below 0.05 mg/kg (the MRL of EU). Results of the ultimate residues in garlic and soil showed that this pesticide is safe to be used under the recommended dosages.
Sigma-Aldrich
1-Naphthylacetic acid, BioReagent, 1 mg/mL, suitable for plant cell culture
Supelco
1-Naphthaleneacetic acid, PESTANAL®, analytical standard
Sigma-Aldrich
1-Naphthaleneacetic acid, suitable for plant cell culture, BioReagent, ≥95%, crystalline
Sigma-Aldrich
1-Naphthaleneacetic acid, technical grade