Merck
  • Ethyl pyruvate protects rats from phosgene-induced pulmonary edema by inhibiting cyclooxygenase2 and inducible nitric oxide synthase expression.
Ethyl pyruvate protects rats from phosgene-induced pulmonary edema by inhibiting cyclooxygenase2 and inducible nitric oxide synthase expression.
Journal of applied toxicology : JAT (2011-08-06)
Hong-li Chen, Hua Bai, Miao-miao Xi, Riu Liu, Xu-jun Qin, Xin Liang, Wei Zhang, Xiao-di Zhang, Wen-li Li, Chun-xu Hai
ABSTRACT
Phosgene is a poorly water-soluble gas penetrating the lower respiratory tract which can induce acute lung injury characterized by a latent phase of fatal pulmonary edema. Pulmonary edema caused by phosgene is believed to be a consequence of oxidative stress and inflammatory responses. Ethyl pyruvate (EP) has been demonstrated to have anti-inflammatory and anti-oxidative properties in vivo and in vitro. The potential therapeutic role of EP in phosgene-induced pulmonary edema has not been addressed so far. In the present study, we aim to investigate the protective effects of EP on phosgene-induced pulmonary edema and the underlying mechanisms. Rats were administered with EP (40 mg kg(-1)) and RAW264.7 cells were also incubated with it (0, 2, 5 or 10 µm) immediately after phosgene (400 ppm, 1 min) or air exposure. Wet-to-dry lung weight ratio (W:D ratio), nitric oxide (NO) and prostaglandin E(2) (PGE(2)) production, cyclooxygenase2 (COX-2) and inducible nitric oxide synthase (iNOS) expression, and mitogen-activated protein kinases activities (MAPKs) were measured. Our results showed that EP treatment attenuated phosgene-induced pulmonary edema and decreased the level of NO and PGE(2) dose-dependently. Furthermore, EP significantly reduced COX-2 expression, iNOS expression and MAPK activation induced by phosgene. Moreover, specific inhibitors of MAPKs reduced COX-2 and iNOS expression induced by phosgene. These findings suggested that EP has a protective role against phosgene-induced pulmonary edema, which is mediated in part by inhibiting MAPK activation and subsequently down-regulating COX-2 and iNOS expression as well as decreasing the production of NO and PGE(2).
Sigma-Aldrich
Phosgene solution, 15 wt. % in toluene
Sigma-Aldrich
Phosgene solution, ~20% in toluene