Merck
  • Genetic variation in base excision repair pathway genes, pesticide exposure, and prostate cancer risk.
Genetic variation in base excision repair pathway genes, pesticide exposure, and prostate cancer risk.
Environmental health perspectives (2011-08-04)
Kathryn Hughes Barry, Stella Koutros, Sonja I Berndt, Gabriella Andreotti, Jane A Hoppin, Dale P Sandler, Laurie A Burdette, Meredith Yeager, Laura E Beane Freeman, Jay H Lubin, Xiaomei Ma, Tongzhang Zheng, Michael C R Alavanja
ABSTRACT
Previous research indicates increased prostate cancer risk for pesticide applicators and pesticide manufacturing workers. Although underlying mechanisms are unknown, evidence suggests a role of oxidative DNA damage. Because base excision repair (BER) is the predominant pathway involved in repairing oxidative damage, we evaluated interactions between 39 pesticides and 394 tag single-nucleotide polymorphisms (SNPs) for 31 BER genes among 776 prostate cancer cases and 1,444 male controls in a nested case-control study of white Agricultural Health Study (AHS) pesticide applicators. We used likelihood ratio tests from logistic regression models to determine p-values for interactions between three-level pesticide exposure variables (none/low/high) and SNPs (assuming a dominant model), and the false discovery rate (FDR) multiple comparison adjustment approach. The interaction between fonofos and rs1983132 in NEIL3 [nei endonuclease VIII-like 3 (Escherichia coli)], which encodes a glycosylase that can initiate BER, was the most significant overall [interaction p-value (pinteract) = 9.3 × 10-6; FDR-adjusted p-value = 0.01]. Fonofos exposure was associated with a monotonic increase in prostate cancer risk among men with CT/TT genotypes for rs1983132 [odds ratios (95% confidence intervals) for low and high use compared with no use were 1.65 (0.91, 3.01) and 3.25 (1.78, 5.92), respectively], whereas fonofos was not associated with prostate cancer risk among men with the CC genotype. Carbofuran and S-ethyl dipropylthiocarbamate (EPTC) interacted similarly with rs1983132; however, these interactions did not meet an FDR < 0.2. Our significant finding regarding fonofos is consistent with previous AHS findings of increased prostate cancer risk with fonofos exposure among those with a family history of prostate cancer. Although requiring replication, our findings suggest a role of BER genetic variation in pesticide-associated prostate cancer risk.