Merck
  • Nanoscale domains with nematic order in supercooled vitamin-A acetate: molecular dynamics studies.
Nanoscale domains with nematic order in supercooled vitamin-A acetate: molecular dynamics studies.
Physical review. E, Statistical, nonlinear, and soft matter physics (2011-07-07)
Z Wojnarowska, M Paluch, P Wlodarczyk, L Hawelek, R Wrzalik, J Zioło, M Wygledowska-Kania, B Bergler-Czop, L Brzezinska-Wcislo, P Bujak
ABSTRACT
Vitamin-A acetate is one of the most versatile vitamins. It is applied in medicine because of its antioxidative properties, in tumor therapy because of its cytostatic activity, and in cosmetics because of its nutritional additives. Herein, using broadband dielectric spectroscopy, the molecular dynamics of supercooled and glassy vitamin-A acetate was investigated. It was shown that dielectric measurements carried out at ambient and elevated pressures reveal a number of relaxation processes associated with different types of molecular motions: α, δ, and ν processes-observed above the glass transition temperature and the next two modes: β and γ identified in the glassy state. The occurrence of the δ mode in the dielectric spectrum may imply the existence of nanoscale domains with nematic order. This hypothesis is further checked by atomic force microscopy measurements. Finally, we have determined the value of the glass transition temperature (T(g)) as well as the steepness index (m(P)) at various T-P conditions.
Sigma-Aldrich
Retinyl acetate, solid or viscous liquid, BioReagent, synthetic, suitable for cell culture
Sigma-Aldrich
Retinyl acetate, synthetic, matrix dispersion, 475,000-650,000 USP units/g
Retinol acetate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Retinyl acetate, synthetic, crystalline solid or supercooled liquid
Supelco
Retinyl acetate, analytical standard
Supelco
Retinyl Acetate (Vitamin A Acetate), Pharmaceutical Secondary Standard; Certified Reference Material