Merck
Biological pH buffers in IVF: help or hindrance to success.
Journal of assisted reproduction and genetics (2011-05-27)
Matthew A Will, Natalie A Clark, Jason E Swain
ABSTRACT
Minimizing environmental stress helps maintain cellular homeostasis and is a crucial component in optimizing embryo development in vitro and resulting ART success. One stressor of particular interest is pH. Biologic buffers, such as HEPES and MOPS, are valuable tools for stabilizing pH. The objective of this manuscript is to summarize efficacy and impact of various pH buffers used during IVF lab procedures Keyword searches were performed using Pubmed and Medline and relevant literature reviewed. Various pH buffers have been used with varying degrees of success for gamete and embryo processing in a variety of animal species, as well as in human. Though biologic buffers off a means to improve pH stability, not all buffers may be appropriate for use with gametes and embryos. Specific buffers may have undesired effects, and these may be buffer, species, cell type or concentration dependent. Continued research is needed to further refine and improve the use of biologic buffers for use in human ART.
Sigma-Aldrich
HEPES sodium salt solution, BioReagent, 1M, suitable for cell culture
SAFC
HEPES sodium salt
Sigma-Aldrich
MOPS, BioUltra, for molecular biology, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES sodium salt, ≥99.0% (titration)
Sigma-Aldrich
HEPES sodium salt, BioPerformance Certified, suitable for cell culture, ≥99.0%
Sigma-Aldrich
MOPS, BioXtra, ≥99.5% (titration)
Sigma-Aldrich
MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)
Sigma-Aldrich
MOPS, ≥99.5% (titration)
SAFC
HEPES
Sigma-Aldrich
HEPES sodium salt, ≥99.5% (titration), free-flowing, Redi-Dri
Sigma-Aldrich
HEPES solution, BioPerformance Certified, 1 M, suitable for cell culture, 0.2 μm filtered
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
HEPES solution, 1 M, pH 7.0-7.6, sterile-filtered, BioReagent, suitable for cell culture