Merck
  • Acetyl chloride-silver nitrate-acetonitrile: a reagent system for the synthesis of 2-nitroglycals and 2-nitro-1-acetamido sugars from glycals.
Acetyl chloride-silver nitrate-acetonitrile: a reagent system for the synthesis of 2-nitroglycals and 2-nitro-1-acetamido sugars from glycals.
The Journal of organic chemistry (2011-05-27)
Pavan K Kancharla, Y Suman Reddy, Suresh Dharuman, Yashwant D Vankar
ABSTRACT
A new reagent system comprising acetyl chloride, silver nitrate, and acetonitrile has been developed for the synthesis of 2-nitroglycals from the corresponding glycals. Under certain conditions, the formation of 2-nitro-1-acetamido sugars has also been observed. In addition, a few other non-carbohydrate-derived olefins also gave the corrresponding conjugated nitroolefins.
Sigma-Aldrich
Acetyl chloride, reagent grade, 98%
Sigma-Aldrich
Acetyl chloride, reagent grade, 98%
Sigma-Aldrich
Acetyl chloride, puriss. p.a., ≥99.0% (T)