- Effects of the organophosphate insecticide azinphos-methyl on the reproduction and cholinesterase activity of Biomphalaria glabrata.
Effects of the organophosphate insecticide azinphos-methyl on the reproduction and cholinesterase activity of Biomphalaria glabrata.
Azinphos-methyl is an organophosphate insecticide used for pest control on a number of food crops in many parts of the world. The snail Biomphalaria glabrata is a freshwater gastropod widely distributed in South America, Central America and Africa. The aim of the present work was to investigate whether azinphos-methyl causes alterations in the reproduction of B. glabrata. To this end, gastropod pigmented specimens were exposed to various concentrations of the insecticide (0.021, 0.5, 2.5, and 5 mg L(-1)) for either 2 or 14 d. Along 14 d, several reproduction parameters and cholinesterase (ChE) activity were evaluated. In each group, the number of egg masses, the number of eggs per mass, the number of hatchings, the time to hatching, and the survival of the offspring after one month of treatment was evaluated. The results showed that, depending on the concentration and time of exposure, azinphos-methyl induced alterations in the reproduction of B. glabrata. These alterations were mainly represented by a decrease in the number of egg masses, and, in some cases, by a lower number or even the total absence of hatchings. Thus, the gastropods exposed to 2.5 and 5 mg L(-1) of azinphos-methyl for 14 d showed ChE inhibitions higher than 35% along time and completely lost their ability to reproduce. On the other hand, exposure to high acute concentrations or exposure to low concentrations for 14 d resulted in ChE inhibition equal to or lower than 35% between 7 and 14 d of treatment and similar alterations in reproduction. These were represented by a decrease in the number of egg masses. At low pestice levels, the number of egg masses and the number of offspring resulted to be more sensitive biomarkers than ChE inhibition. It is concluded that the insecticide azinphos-methyl can cause a decline in the reproductive performance of B. glabrata.