Merck
  • Chemical absorption of carbon dioxide with asymmetrically heated polytetrafluoroethylene membranes.
Chemical absorption of carbon dioxide with asymmetrically heated polytetrafluoroethylene membranes.
Journal of environmental management (2011-01-05)
Shia-Chung Chen, Su-Hsia Lin, Rean-Der Chien, Yuen-Hua Wang, Hseun-Chi Hsiao
ABSTRACT
In this study, the absorption of carbon dioxide using an absorbent composed of 2-amino-2-methyl-L-propanol (AMP) + monoethanolamine (MEA) + piperazine (PZ) in asymmetric and symmetric polytetrafluoroethylene (PTFE) membrane contactors was investigated. Experiments were conducted using various gas flow rates, liquid flow rates, and absorbent blends. CO(2) recovery increased with increasing liquid flow rates. The mean pore size of PTFE membrane reduced via heating treatment. An asymmetric membrane had a better CO(2) recovery than a symmetric membrane. For the asymmetric membrane, placing the smaller pore-size side of the membrane in contact with the liquid phase, reduced the level of wetting of the membrane. The membrane mass transfer coefficient and durability of the PTFE membrane were enhanced by asymmetrically heating.
Sigma-Aldrich
2-Amino-2-methyl-1-propanol, ~5% Water, technical grade, 95%
Sigma-Aldrich
2-Amino-2-methyl-1-propanol, BioUltra, ≥99.0% (GC)
Sigma-Aldrich
2-Amino-2-methyl-1-propanol, BioXtra, ≥95%
Sigma-Aldrich
2-Amino-2-methyl-1-propanol, technical, ≥90% (GC)