Merck
  • Cooperative binding and stabilization of the medicinal pigment curcumin by diamide linked γ-cyclodextrin dimers: a spectroscopic characterization.
Cooperative binding and stabilization of the medicinal pigment curcumin by diamide linked γ-cyclodextrin dimers: a spectroscopic characterization.
The journal of physical chemistry. B (2011-01-05)
Takaaki Harada, Duc-Truc Pham, Mandy H M Leung, Huy Tien Ngo, Stephen F Lincoln, Christopher J Easton, Tak W Kee
ABSTRACT
Diamide linked γ-cyclodextrin (γ-CD) dimers are used to capture curcumin and suppress its decomposition in water. In this study, succinamide and urea linked γ-CD dimers joined through the C6(A) carbon on each γ-CD are used. The γ-CD dimers, 66γCD(2)su and 66γCD(2)ur, show a remarkable ability to suppress the decomposition of curcumin and extend its half-life from less than 30 min to greater than 16 h. The 1:1 association of curcumin with 66γCD(2)su and 66γCD(2)ur has high stability constants of 8.7 × 10(6) M(-1) and 2.0 × 10(6) M(-1), respectively. In addition, 2D (1)H NOESY NMR results show specific hydrogen interactions in the association of curcumin with 66γCD(2)su and 66γCD(2)ur, consistent with the cooperative binding of curcumin by both γ-CD annuli of 66γCD(2)su and 66γCD(2)ur. The interactions between curcumin in the linked γ-CD dimers and surfactant micelles were studied using fluorescence spectroscopy. While linked γ-CD dimer-bound curcumin has a negligible fluorescence quantum yield, a significant increase in fluorescence intensity (Φ(fl) > 2%) in the presence of micelles suggests that curcumin is delivered to the micelle. The overall results indicate that the diamide linked γ-CD dimers are highly promising systems for curcumin delivery in vivo due to effective curcumin stabilization.
Sigma-Aldrich
γ-Cyclodextrin, powder, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
γ-Cyclodextrin, Wacker Chemie AG, ≥90.0% cyclodextrin basis (HPLC)
Sigma-Aldrich
γ-Cyclodextrin, ≥98%