Merck
Binding of more than one retinoid to visual opsins.
Biophysical journal (2010-10-07)
Clint L Makino, Charles K Riley, James Looney, Rosalie K Crouch, Tetsuji Okada
ABSTRACT
Visual opsins bind 11-cis retinal at an orthosteric site to form rhodopsins but increasing evidence suggests that at least some are capable of binding an additional retinoid(s) at a separate, allosteric site(s). Microspectrophotometric measurements on isolated, dark-adapted, salamander photoreceptors indicated that the truncated retinal analog, β-ionone, partitioned into the membranes of green-sensitive rods; however, in blue-sensitive rod outer segments, there was an enhanced uptake of four or more β-ionones per rhodopsin. X-ray crystallography revealed binding of one β-ionone to bovine green-sensitive rod rhodopsin. Cocrystallization only succeeded with extremely high concentrations of β-ionone and binding did not alter the structure of rhodopsin from the inactive state. Salamander green-sensitive rod rhodopsin is also expected to bind β-ionone at sufficiently high concentrations because the binding site is present on its surface. Therefore, both blue- and green-sensitive rod rhodopsins have at least one allosteric binding site for retinoid, but β-ionone binds to the latter type of rhodopsin with low affinity and low efficacy.
Sigma-Aldrich
β-Ionone, natural, ≥95%, FG
Sigma-Aldrich
β-Ionone, 96%
Sigma-Aldrich
β-Ionone, predominantly trans, ≥97%, FCC, FG