Merck
Sulfhydryl-based dendritic chain reaction.
Chemical communications (Cambridge, England) (2010-08-18)
Eran Sella, Roy Weinstain, Rotem Erez, Noah Z Burns, Phil S Baran, Doron Shabat
ABSTRACT
A new dendritic chain reaction probe system was demonstrated to produce exponential signal amplification for the detection of sulfhydryl compounds.
Sigma-Aldrich
5-Amino-2-nitrobenzoic acid, 97%