Merck
  • Biological activity of modified and exchanged 2-amino-5-nitrothiazole amide analogues of nitazoxanide.
Biological activity of modified and exchanged 2-amino-5-nitrothiazole amide analogues of nitazoxanide.
Bioorganic & medicinal chemistry letters (2010-05-22)
T Eric Ballard, Xia Wang, Igor Olekhnovich, Taylor Koerner, Craig Seymour, Paul S Hoffman, Timothy L Macdonald
ABSTRACT
Head group analogues of the antibacterial and antiparasitic drug nitazoxanide (NTZ) are presented. A library of 39 analogues was synthesized and assayed for their ability to suppress growth of Helicobacter pylori, Campylobacter jejuni, Clostridium difficile and inhibit NTZ target pyruvate:ferredoxin oxidoreductase (PFOR). Two head groups assayed recapitulated NTZ activity and possessed improved activity over their 2-amino-5-nitrothiazole counterparts, demonstrating that head group modification is a viable route for the synthesis of NTZ-related antibacterial analogues.
Sigma-Aldrich
2-Amino-5-nitrothiazole, 97%