Merck
  • Induction or inhibition of cytochrome P450 2E1 modifies the acute toxicity of acrylonitrile in rats: biochemical evidence.
Induction or inhibition of cytochrome P450 2E1 modifies the acute toxicity of acrylonitrile in rats: biochemical evidence.
Archives of toxicology (2010-02-04)
Wang Suhua, Lu Rongzhu, Xu Wenrong, Xing Guangwei, Zhao Xiaowu, Wang Shizhong, Zhang Ye, Han Fangan, Michael Aschner
ABSTRACT
The present study was designed to examine the effects of the inhibition or induction of CYP2E1 activity on acute acrylonitrile (AN) toxicity in rats. Increased or decreased hepatic CYP2E1 activity was achieved by pretreatment with acetone or trans-1,2-dichloroethylene (DCE), respectively. AN (50 mg/kg) was administered by intraperitoneal injection. Onset of convulsions and death were observed in rats with increased CYP2E1 activity, whereas convulsions and death did not appear in rats within 1 h after treatment with AN alone. Convulsions occurred in all AN-treated animals with increased CYP2E1 activity at approximately 18 min. The levels of cyanide (CN(-)), a terminal metabolite of AN, were significantly increased in the brains and livers of the AN-treated rats with increased CYP2E1 activity, compared with the levels in rats treated with AN alone, DCE + AN or acetone + DCE + AN. The cytochrome c oxidase (CcOx) activities in the brains and livers of the rats treated with AN or AN + acetone were significantly lower than those in the normal control rats and the rats treated with DCE, whereas the CcOx activities in the brains and livers of rats with decreased CYP2E1 activity were significantly higher than those in AN-treated rats. Brain lipid peroxidation was enhanced, and the antioxidant capacity was significantly compromised in rats with decreased CYP2E1 activity compared with rats with normal or increased CYP2E1 activity. Therefore, inhibition of CYP2E1 and simultaneous antioxidant therapy should be considered as supplementary therapeutic interventions in acute AN intoxication cases with higher CYP2E1 activity, thus a longer window of opportunity would be got to offer further emergency medication.
Product Description
Supelco
cis-1,2-Dichloroethene solution, certified reference material, 2000 μg/mL in methanol
Supelco
trans-1,2-Dichloroethylene, analytical standard
Sigma-Aldrich
1,2-Dichloroethylene, mixture of cis and trans, 98%
Sigma-Aldrich
cis-1,2-Dichloroethylene, 97%
Sigma-Aldrich
trans-1,2-Dichloroethylene, 98%
Supelco
cis-1,2-Dichloroethene, analytical standard