Merck
  • Optical waveguide sensor of volatile organic compounds based on PTA thin film.
Optical waveguide sensor of volatile organic compounds based on PTA thin film.
Analytica chimica acta (2010-01-20)
Renagul Abdurahman, Abliz Yimit, Hayrensa Ablat, Mamtimin Mahmut, Ji De Wang, Kiminori Itoh
ABSTRACT
In this study, a sensitive optical waveguide (OWG) sensor for the detection and identification of volatile organic compounds (VOCs) was reported. The sensing membrane is constructed by immobilization of peroxopolytungsten acid (PTA) thin film over a single-mode potassium ion (K(+)) exchanged glass OWG by spin-coating method. A laser beam was coupled into and out of the glass optical waveguide using prism couplers, and dry air functioned as a carrier gas. The sensor was tested for various volatile organic compounds (VOCs), and it showed higher response to the chlorobenzene gas compared to other VOCs. Therefore, we used the OWG sensor to detect chlorobenzene gas as a typical example of VOCs. The sensor exhibits a linear response to chlorobenzene gas in the range of 0.4-1000 ppm with rapid response and good reversibility. The constructed sensor is easy to fabricate and it has some unique qualities which can be characterized as inexpensive, sensitive, and reusable.
Sigma-Aldrich
Tungstic acid, 99%
Sigma-Aldrich
Tungstic acid, ≥98.0% (calcined substance, T), powder