Merck
  • [(18)F]-labeled 2-methoxyphenylpiperazine derivative as a potential brain positron emission tomography imaging agent.
[(18)F]-labeled 2-methoxyphenylpiperazine derivative as a potential brain positron emission tomography imaging agent.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine (2009-08-15)
Tiantian Mou, Wenjiang Yang, Cheng Peng, Xianzhong Zhang, Yunchuan Ma
ABSTRACT
This study reports the synthesis and characterization of N-(3-(4-(2-methoxyphenyl)piperazin-1-yl)propyl-4-[(18)F]fluorobenzamide ([(18)F]MPP3F). The total reaction time for [(18)F]MPP3F, including final high-performance liquid chromatography purification, was about 3h. Typical decay-corrected radiochemical yield was 18.4+/-3.1%. The radiochemical purity was >98%. Biodistribution in mice showed that [(18)F]MPP3F is a potential brain imaging agent for positron emission tomography. The brain uptake of [(18)F]MPP3F was 6.59+/-0.77% Injected Dose/g at 2 min post-injection time. A brain-to-blood ratio of 3.67 was reached at 15 min after injection.